CAS RN 37088-27-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-5-(dimethylsulfamoyl)benzoic Acid, TRC
CAS: 37088-27-0 Molecular Formula: C9H10NO4SCl Molecular Weight (g/mol): 263.7 IUPAC Name: 2-chloro-5-(dimethylsulfamoyl)benzoic acid SMILES: CN(C)S(=O)(=O)c1ccc(Cl)c(c1)C(=O)O