accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNClH3CCl

CAS RN 37091-73-9

IUPAC Name: 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium chloride
Synonyms: 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride
Molecular Weight (g/mol): 169.05
Molecular Formula: C5H10Cl2N2
InChi Key: AEBBXVHGVADBHA-UHFFFAOYSA-M
SMILES: [Cl-].CN1CC[N+](C)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results