accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONH2

CAS RN 371-34-6

IUPAC Name: ethyl 6-aminohexanoate
Synonyms: ethyl 6-aminohexanoate
Molecular Weight (g/mol): 159.23
Molecular Formula: C8H17NO2
InChi Key: NJNQDCIAOXIFTB-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results