accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 371-40-4

IUPAC Name: 4-fluoroaniline
Synonyms: 4-fluoroaniline
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H6FN
InChi Key: KRZCOLNOCZKSDF-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)