accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2CNHOHNO

CAS RN 37107-81-6

IUPAC Name: 5-ethenyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-ethenyl-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: ZRYZBEQILKESAW-UHFFFAOYSA-N
SMILES: C=CC1=CNC(=O)NC1=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)