accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37112-31-5

IUPAC Name: (1S,5R)-6,8-dioxabicyclo[3.2.1]oct-2-en-4-one
Synonyms: levoglucosenone
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: HITOXZPZGPXYHY-UJURSFKZSA-N
SMILES: O=C1C=C[C@H]2CO[C@@H]1O2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results