accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37128-24-8

IUPAC Name: ethyl 2-(2-methyl-1,3-thiazol-4-yl)acetate
Synonyms: ethyl 2-(2-methyl-1,3-thiazol-4-yl)acetate
Molecular Weight (g/mol): 185.24
Molecular Formula: C8H11NO2S
InChi Key: DWSMURKWFWKPBT-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CSC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results