CAS RN 371753-19-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac trans-Loxoprofen Alcohol, TRC
CAS: 371753-19-4 Molecular Formula: C15H20O3 Molecular Weight (g/mol): 248.32 Synonym: rel-4-[[(1R,2S)-2-Hydroxycyclopentyl]methyl]-α-methylbenzeneacetic Acid; IUPAC Name: 2-[4-[[(1R,2S)-2-hydroxycyclopentyl]methyl]phenyl]propanoic acid SMILES: O[C@H]1CCC[C@@H]1CC2=CC=C(C(C)C(O)=O)C=C2