accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 371766-08-4

IUPAC Name: (isoquinolin-5-yl)boronic acid
Synonyms: isoquinolin-5-ylboronic acid
Molecular Weight (g/mol): 172.98
Molecular Formula: C9H8BNO2
InChi Key: XKEYHBLSCGBBGU-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CN=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results