accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 372-09-8

IUPAC Name: 2-cyanoacetic acid
Synonyms: cyanoacetic acid cyanessigsaeure acide cyanacetique malonic acid mononitrile malonic mononitrile
Molecular Weight (g/mol): 85.06
Molecular Formula: C3H3NO2
InChi Key: MLIREBYILWEBDM-UHFFFAOYSA-N
SMILES: OC(=O)CC#N
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (4)