accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOHFFF

CAS RN 372-30-5

IUPAC Name: ethyl 4,4,4-trifluoro-3-hydroxybutanoate
Synonyms: ethyl 4,4,4-trifluoro-3-hydroxybutanoate
Molecular Weight (g/mol): 186.13
Molecular Formula: C6H9F3O3
InChi Key: ZWEDFBKLJILTMC-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results