accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOO

CAS RN 3721-95-7

IUPAC Name: cyclobutanecarboxylic acid
Synonyms: cyclobutanecarboxylic acid
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: TXWOGHSRPAYOML-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC1
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results