accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NONH2

CAS RN 3724-16-1

IUPAC Name: 2-(pyridin-3-yl)acetamide
Synonyms: 3-pyridine acetamide
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: YDHIMEXEGOCNHU-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results