accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHPOOPOHOOPHOOO

CAS RN 37267-86-0

IUPAC Name: trihydroxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-trione
Synonyms: trimetaphosphoric acid magnesium (2:3) salt trimetaphosphoric acid trisodium salt trimetaphosphoric acid magnesium trimetaphosphate calcium trimetaphosphate trimetaphosphoric acid, tripotassium salt metaphosphoric acid ruthenium (+3) salt trimetaphosphoric acid trimetaphosphoric acid, ruthenium (+3) salt trimetaphosphoric acid, magnesium (2:3) salt
Molecular Weight (g/mol): 239.94
Molecular Formula: H3O9P3
InChi Key: AZSFNUJOCKMOGB-UHFFFAOYSA-N
SMILES: OP1(=O)OP(O)(=O)OP(O)(=O)O1
Quantity 
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (1)
  • (1)
  • (8)
  • (1)
Grade 
  • (3)
  • (8)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (9)
  • (3)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (1)
  • (1)
  • (8)
  • (1)
  • (3)
  • (8)
  • (2)
  • (2)
  • (2)
  • (2)