accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 373-61-5

OH3CHOOCH3OHFBFF

CAS RN 373-61-5

IUPAC Name: boron(3+) dihydrogen diacetate trifluoride
Synonyms: boron(3+) dihydrogen diacetate trifluoride
Molecular Weight (g/mol): 187.91
Molecular Formula: C4H8BF3O4
InChi Key: OVYKFNOIPQYMRU-UHFFFAOYSA-K
SMILES: [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)