accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOHFF

CAS RN 373-96-6

IUPAC Name: 2,2-difluoropropanoic acid
Synonyms: 2,2-difluoropropanoic acid
Molecular Weight (g/mol): 110.06
Molecular Formula: C3H4F2O2
InChi Key: PMWGIVRHUIAIII-UHFFFAOYSA-N
SMILES: CC(F)(F)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results