CAS RN 374-25-4
IUPAC Name:
4-bromo-3-chloro-3,4,4-trifluorobut-1-ene
Synonyms:
4-bromo-3-chloro-3,4,4-trifluorobut-1-ene
Molecular Weight (g/mol):
223.42
Molecular Formula:
C4H3BrClF3
InChi Key:
VULPFOSLGWWARI-UHFFFAOYNA-N
SMILES:
FC(F)(Br)C(F)(Cl)C=C