accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 374-35-6

CH3OHOFFFHO

CAS RN 374-35-6

IUPAC Name: 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
Synonyms: 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
Molecular Weight (g/mol): 158.08
Molecular Formula: C4H5F3O3
InChi Key: CTGJACFEVDCYMC-UHFFFAOYNA-N
SMILES: CC(O)(C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results