accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 374-99-2

IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutan-1-aminium
Synonyms: 2,2,3,3,4,4,4-heptafluorobutan-1-aminium
Molecular Weight (g/mol): 200.08
Molecular Formula: C4H5F7N
InChi Key: WBGBQSRNXPVFDB-UHFFFAOYSA-O
SMILES: [NH3+]CC(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results