accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONOO

CAS RN 3740-52-1

IUPAC Name: 2-(2-nitrophenyl)acetic acid
Synonyms: (2-nitrophenyl)acetic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: WMUZDBZPDLHUMW-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)