accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NONHBr

CAS RN 374063-92-0

IUPAC Name: 5-bromo-1,2-dihydropyrazin-2-one
Synonyms: 5-bromo-1H-pyrazin-2-one
Molecular Weight (g/mol): 174.99
Molecular Formula: C4H3BrN2O
InChi Key: ITTXBHQAWOFJAI-UHFFFAOYSA-N
SMILES: BrC1=CNC(=O)C=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results