accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 374068-01-6

IUPAC Name: 2-chloropyrimidine-5-carboxylic acid
Synonyms: 2-chloropyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: DUCXUPKLVVSJKA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(Cl)N=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results