accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 374538-00-8

BONOCH3Br

CAS RN 374538-00-8

IUPAC Name: 2-(3-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
Synonyms: 2-(3-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
Molecular Weight (g/mol): 283.96
Molecular Formula: C11H15BBrNO2
InChi Key: CYGPBFLFZQMUBN-UHFFFAOYSA-N
SMILES: CN1CCOB(OCC1)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results