accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BHOOH

CAS RN 374538-04-2

IUPAC Name: (4-cyclohexylphenyl)boronic acid
Synonyms: 4-cyclohexylphenylboronic acid
Molecular Weight (g/mol): 204.08
Molecular Formula: C12H17BO2
InChi Key: KNQVRFYNQWNYPU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results