accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37466-89-0

IUPAC Name: 3-methoxybenzene-1,2-diamine
Synonyms: 3-methoxybenzene-1,2-diamine
Molecular Weight (g/mol): 138.17
Molecular Formula: C7H10N2O
InChi Key: BFLWXPJTAKXXKT-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results