accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 375-80-4

FFIFFFFFFFFFFI

CAS RN 375-80-4

IUPAC Name: 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
Synonyms: 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
Molecular Weight (g/mol): 553.86
Molecular Formula: C6F12I2
InChi Key: JOQDDLBOAIKFQX-UHFFFAOYSA-N
SMILES: FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)