accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFFFFFFFFFFFFBr

CAS RN 375-88-2

IUPAC Name: 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane
Synonyms: 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane
Molecular Weight (g/mol): 448.96
Molecular Formula: C7BrF15
InChi Key: VPQQZKWYZYVTMU-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)