accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 375-95-1

HOOFFFFFFFFFFFFFFFFF

CAS RN 375-95-1

IUPAC Name: heptadecafluorononanoic acid
Synonyms: perfluorononanoic acid heptadecafluornonansaeure perfluornonansaeure PFNA heptadekafluornonansaeure
Molecular Weight (g/mol): 464.08
Molecular Formula: C9HF17O2
InChi Key: UZUFPBIDKMEQEQ-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results