accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37560-50-2

IUPAC Name: 1-(4-chlorophenyl)-1H-pyrrole-2-carbaldehyde
Synonyms: 1-(4-chlorophenyl)pyrrole-2-carbaldehyde
Molecular Weight (g/mol): 205.64
Molecular Formula: C11H8ClNO
InChi Key: NNQOVJLQFYROBU-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)N1C=CC=C1C=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results