accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNSBrNH2

CAS RN 37566-39-5

IUPAC Name: 5-bromo-1,3,4-thiadiazol-2-amine
Synonyms: 5-bromo-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 180.02
Molecular Formula: C2H2BrN3S
InChi Key: GLYQQFBHCFPEEU-UHFFFAOYSA-N
SMILES: NC1=NN=C(Br)S1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)