CAS RN 375857-64-0
IUPAC Name:
5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
Synonyms:
5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol):
239.07
Molecular Formula:
C9H7BrN2O
InChi Key:
NYAFKUCVAVUCHW-UHFFFAOYSA-N
SMILES:
CC1=NOC(=N1)C1=CC=CC=C1Br
1
–
1
of
1
results
1
5-(2-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™
CAS: 375857-64-0 Molecular Formula: C9H7BrN2O Molecular Weight (g/mol): 239.072 InChI Key: NYAFKUCVAVUCHW-UHFFFAOYSA-N PubChem CID: 18001310 IUPAC Name: 5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole SMILES: CC1=NOC(=N1)C2=CC=CC=C2Br