accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOFFFFFFFFFFFFFFFF

CAS RN 376-18-1

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononan-1-ol
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononan-1-ol
Molecular Weight (g/mol): 432.1
Molecular Formula: C9H4F16O
InChi Key: MSXVQELLSMPBFD-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)