accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3SiH3CCH3NCH3H3C

CAS RN 3768-58-9

IUPAC Name: [(dimethylamino)dimethylsilyl]dimethylamine
Synonyms: [(dimethylamino)dimethylsilyl]dimethylamine
Molecular Weight (g/mol): 146.31
Molecular Formula: C6H18N2Si
InChi Key: QULMGWCCKILBTO-UHFFFAOYSA-N
SMILES: CN(C)[Si](C)(C)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results