accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SHClClCl

CAS RN 3773-14-6

IUPAC Name: 2,4,5-trichlorobenzene-1-thiol
Synonyms: 2,4,5-trichlorothiophenol
Molecular Weight (g/mol): 213.5
Molecular Formula: C6H3Cl3S
InChi Key: JARIALSGFXECCH-UHFFFAOYSA-N
SMILES: SC1=CC(Cl)=C(Cl)C=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results