accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHNPOONCl

CAS RN 3778-73-2

IUPAC Name: 3-(2-chloroethyl)-2-[(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one
Synonyms: ifosfamide isofosfamide iphosphamide holoxan iso endoxan mitoxana ifosfamide (product) iso-endoxan isosfamide IFF
Molecular Weight (g/mol): 261.08
Molecular Formula: C7H15Cl2N2O2P
InChi Key: HOMGKSMUEGBAAB-UHFFFAOYNA-N
SMILES: ClCCNP1(=O)OCCCN1CCCl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Percent Purity 
  • (2)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results