accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOH

CAS RN 3780-50-5

IUPAC Name: 4-(octyloxy)phenol
Synonyms: 4-(octyloxy)phenol
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: HFRUPPHPJRZOCM-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results