accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 378211-85-9

IUPAC Name: pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Synonyms: pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: DZPAOAZDQHZRGG-UHFFFAOYSA-N
SMILES: OC(=O)C1=NN2C=CC=NC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results