accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OCl

CAS RN 37859-25-9

IUPAC Name: 2-(naphthalen-2-yl)acetyl chloride
Synonyms: 2-(naphthalen-2-yl)acetyl chloride
Molecular Weight (g/mol): 204.65
Molecular Formula: C12H9ClO
InChi Key: QEJGMKHQXSZCOS-UHFFFAOYSA-N
SMILES: ClC(=O)CC1=CC2=CC=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results