accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 37888-03-2

ClNOO(E/Z)

CAS RN 37888-03-2

IUPAC Name: 1-chloro-3-(2-nitroethenyl)benzene
Synonyms: 3-chloro-B-nitrostyrene
Molecular Weight (g/mol): 183.59
Molecular Formula: C8H6ClNO2
InChi Key: GXQRAWTWDNHGBS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C=CC1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results