accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOCH3Cl

CAS RN 37908-98-8

IUPAC Name: methyl 3-chloro-4-methoxybenzoate
Synonyms: methyl 3-chloro-4-methoxybenzoate
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: PINQDVFQCCFACD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results