accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3OONH2BOHHO

CAS RN 380430-56-8

IUPAC Name: hydrogen [3-amino-5-(methoxycarbonyl)phenyl]boronic acid chloride
Synonyms: hydrogen 3-amino-5-(methoxycarbonyl)phenylboronic acid chloride
Molecular Weight (g/mol): 231.44
Molecular Formula: C8H11BClNO4
InChi Key: NOMZZWSCDMMVAF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=O)C1=CC(N)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results