accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 380906-80-9

ClOOClCl

CAS RN 380906-80-9

IUPAC Name: 5-(2,5-dichlorophenyl)furan-2-carbonyl chloride
Synonyms: 5-(2,5-dichlorophenyl)furan-2-carbonyl chloride
Molecular Weight (g/mol): 275.51
Molecular Formula: C11H5Cl3O2
InChi Key: BSRRSYCAENNRPG-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(O1)C1=C(Cl)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results