accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOFF

CAS RN 381-73-7

IUPAC Name: 2,2-difluoroacetic acid
Synonyms: difluoroacetic acid copper (+2) salt difluoroacetic acid difluoroacetic acid, lead (+4) salt lead (+4) salt difluoroacetic acid DFA difluoroacetic acid, copper (+1) salt ammonium salt difluoroacetic acid sodium salt difluoroacetic acid silver (+1) salt difluoroacetic acid difluoroacetic acid, silver (+1) salt
Molecular Weight (g/mol): 96.03
Molecular Formula: C2H2F2O2
InChi Key: PBWZKZYHONABLN-UHFFFAOYSA-N
SMILES: OC(=O)C(F)F
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results