accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CBr

CAS RN 3814-34-4

IUPAC Name: 3-(bromomethyl)pentane
Synonyms: 3-(bromomethyl)pentane
Molecular Weight (g/mol): 165.07
Molecular Formula: C6H13Br
InChi Key: KKGUMGWNFARLSL-UHFFFAOYSA-N
SMILES: CCC(CC)CBr
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)