accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONNH

CAS RN 38212-30-5

IUPAC Name: 1-(4-methoxyphenyl)piperazine
Synonyms: para-methoxyphenylpiperazine
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: MRDGZSKYFPGAKP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)