accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOOOOOH2OH2OH2OH2ODyDy

CAS RN 38245-35-1

IUPAC Name: didysprosium(3+) tetrahydrate tricarbonate
Synonyms: didysprosium(3+) tetrahydrate tricarbonate
Molecular Weight (g/mol): 577.08
Molecular Formula: C3H8Dy2O13
InChi Key: BQRZSEZQNZPPPU-UHFFFAOYSA-H
SMILES: O.O.O.O.[Dy+3].[Dy+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results