CAS RN 38245-37-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)
CAS: 38245-37-3 Molecular Formula: C3O9Sm2 Molecular Weight (g/mol): 480.74 MDL Number: MFCD00150444 InChI Key: QCZFMLDHLOYOQJ-UHFFFAOYSA-H IUPAC Name: disamarium(3+) tricarbonate SMILES: [Sm+3].[Sm+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O