accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3827-49-4

IUPAC Name: 2-chloro-5-fluorophenol
Synonyms: 2-chloro-5-fluorophenol
Molecular Weight (g/mol): 146.55
Molecular Formula: C6H4ClFO
InChi Key: CMQOZIKIOASEIN-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)