accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNBr

CAS RN 38275-57-9

IUPAC Name: 5-bromopyrimidine-2-carbonitrile
Synonyms: 5-bromopyrimidine-2-carbonitrile
Molecular Weight (g/mol): 184
Molecular Formula: C5H2BrN3
InChi Key: VPQICCOHFSGBMA-UHFFFAOYSA-N
SMILES: BrC1=CN=C(N=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results