accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOFFF

CAS RN 383-63-1

IUPAC Name: ethyl 2,2,2-trifluoroacetate
Synonyms: trifluoroethyl acetate
Molecular Weight (g/mol): 142.08
Molecular Formula: C4H5F3O2
InChi Key: STSCVKRWJPWALQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (4)